Learn More
Description
Specifications
Specifications
| Antigen | CCL2/MCP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | Western Blot 0.5 ug/ml |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF |
| Gene Symbols | CCL2 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide made to an internal portion of rat MCP1 (within residues 30-80). [Swiss-Prot# P14844] |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
