Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL2/MCP1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NBP107034F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP107034F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP107034F Supplier Novus Biologicals Supplier No. NBP107034F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCL2/MCP1 Polyclonal specifically detects CCL2/MCP1 in Human, Rat samples. It is validated for Immunohistochemistry (Negative), ICC/IF (Negative).

Specifications

Antigen CCL2/MCP1
Applications Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry (Negative), ICC/IF (Negative)
Formulation PBS with 0.05% Sodium Azide
Gene Alias C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF
Gene Symbols CCL2
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of rat MCP1 (within residues 30-80). [Swiss-Prot# P14844]
Molecular Weight of Antigen 16 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Asthma, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6347
Test Specificity Human and Rat. Mouse reactivity reported by a customer review. Immunogen sequence has 95% homology to mouse.
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.