Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL2/MCP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP179938 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP179938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP179938 Supplier Novus Biologicals Supplier No. NBP179938
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCL2/MCP1 Polyclonal specifically detects CCL2/MCP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CCL2/MCP1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_002973
Gene Alias C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF
Gene Symbols CCL2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CCL2The immunogen for this antibody is CCL2. Peptide sequence ASYRRITSSKCPKEAVIFKTIVAKEICADPKQKWVQDSMDHLDKQTQTPK.
Molecular Weight of Antigen 11 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6347
Reconstitution Centrifuge prior to opening. Reconstitute with sterilized PBS to a final concentration of 1mg/ml. Vortex followed by Centrifuge again to pellet the solution.
Target Species Human, Canine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.