Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL20/MIP-3 alpha Antibody (114906), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB302706 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302706 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302706 Supplier Novus Biologicals Supplier No. FAB760L
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCL20/MIP-3 alpha Monoclonal specifically detects CCL20/MIP-3 alpha in Mouse samples. It is validated for Western Blot.

Specifications

Antigen CCL20/MIP-3 alpha
Applications Western Blot
Classification Monoclonal
Clone 114906
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL20/MIP-3 alpha , Ala28-Met97, Accession # O89093
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6364
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.