Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL20/MIP-3 alpha Antibody (4N5F7), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP227142Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227142Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227142Z Supplier Novus Biologicals Supplier No. NBP227142AF700
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCL20/MIP-3 alpha Monoclonal specifically detects CCL20/MIP-3 alpha in Human samples. It is validated for Western Blot.

Specifications

Antigen CCL20/MIP-3 alpha
Applications Western Blot
Classification Monoclonal
Clone 4N5F7
Conjugate Alexa Fluor 700
Dilution Western Blot
Gene Alias beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38
Gene Symbols CCL20
Host Species Rat
Immunogen Full length recombinant mouse MIP-3 full length protein
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6364
Test Specificity This monoclonal antibody is specific to Mouse MIP-3 it will not cross-react with Human MIP-3.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.