Learn More
Description
Specifications
Specifications
| Antigen | CCL20/MIP-3 alpha |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 4N5F7 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1-2ug/ml |
| Gene Alias | beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 |
| Gene Symbols | CCL20 |
| Host Species | Rat |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
