Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL20/MIP-3 alpha Antibody (4N5F7) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $426.00
Specifications
| Antigen | CCL20/MIP-3 alpha |
|---|---|
| Clone | 4N5F7 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.1-2ug/ml |
| Applications | Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227142SS
![]() |
Novus Biologicals
NBP227142SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227142
![]() |
Novus Biologicals
NBP227142 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
CCL20/MIP-3 alpha Monoclonal specifically detects CCL20/MIP-3 alpha in Human samples. It is validated for Western Blot.Specifications
| CCL20/MIP-3 alpha | |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Chemokines and Cytokines | |
| beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 | |
| CCL20 | |
| IgG2a κ | |
| Protein G purified | |
| This monoclonal antibody is specific to Mouse MIP-3 it will not cross-react with Human MIP-3. |
| 4N5F7 | |
| Western Blot 0.1-2ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 6364 | |
| Full length recombinant mouse MIP-3 full length protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title