Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL20/MIP-3 alpha Antibody (4N8C7), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP227143A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP227143A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP227143A94 Supplier Novus Biologicals Supplier No. NBP227143AF594

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

CCL20/MIP-3 alpha Monoclonal antibody specifically detects CCL20/MIP-3 alpha in Mouse samples. It is validated for Western Blot.

Specifications

Antigen CCL20/MIP-3 alpha
Applications Western Blot
Classification Monoclonal
Clone 4N8C7
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38
Gene Symbols CCL20
Host Species Rat
Immunogen Full length recombinant mouse CCL20 (MIP-3a) was used as immunogen for the antibody. full length protein
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6364
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.