Learn More
CCL20/MIP-3 alpha Antibody (4N8C7), Alexa Fluor™ 700, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | CCL20/MIP-3 alpha |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 4N8C7 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot |
| Gene Alias | beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 |
| Gene Symbols | CCL20 |
| Host Species | Rat |
| Immunogen | Full length recombinant mouse CCL20 (MIP-3a) was used as immunogen for the antibody. full length protein |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.