Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL21/6Ckine Antibody (59106), FITC, Novus Biologicals™
SDP

Catalog No. NB369793 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369793 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369793 Supplier Novus Biologicals Supplier No. FAB457F
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCL21/6Ckine Monoclonal specifically detects CCL21/6Ckine in Mouse samples. It is validated for CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot.

Specifications

Antigen CCL21/6Ckine
Applications CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 59106
Conjugate FITC
Dilution Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, member 21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLCSecondary lymphoid-tissue chemokine
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL21/6Ckine, Ser24-Gly133, Accession # P84444
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6366
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.