Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL22/MDC Antibody (RM0175-9L23), FITC, Novus Biologicals™
SDP

Catalog No. NB361338 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361338 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361338 Supplier Novus Biologicals Supplier No. NBP212106F
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCL22/MDC Monoclonal antibody specifically detects CCL22/MDC in Mouse samples. It is validated for Western Blot, Neutralization

Specifications

Antigen CCL22/MDC
Applications Western Blot, Neutralization
Classification Monoclonal
Clone RM0175-9L23
Conjugate FITC
Dilution Western Blot, Block/Neutralize
Formulation PBS
Gene Alias A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1
Host Species Rat
Immunogen Mouse recombinant CCL22/MDC
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 6367
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.