Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL23/Ck beta 8-1/MIP3 Antibody (007), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB306933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306933 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306933 Supplier Novus Biologicals Supplier No. NBP289592MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CCL23/Ck beta 8-1/MIP3 Monoclonal antibody specifically detects CCL23/Ck beta 8-1/MIP3 in Human samples. It is validated for ELISA

Specifications

Antigen CCL23/Ck beta 8-1/MIP3
Applications ELISA
Classification Monoclonal
Clone 007
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias C6 beta-chemokine, chemokine (C-C motif) ligand 23, CKb8, CKB-8, CK-beta-8, hmrp-2a, Macrophage inflammatory protein 3, MIP3, MIP-3CK-BETA-8, MPIF1, MPIF-1Ckb-8-1, Myeloid progenitor inhibitory factor 1, SCYA23C-C motif chemokine 23, small inducible cytokine subfamily A (Cys-Cys), member 23, Small-inducible cytokine A23
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL23/Ck beta 8-1/MIP3 (Accession#: AAB51134.1; Arg22-Asn120).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6368
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.