Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL24/Eotaxin-2/MPIF-2 Antibody (61016.11), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB2010370IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB2010370IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB2010370IR Supplier Novus Biologicals Supplier No. NB12010370IR

Mouse Monoclonal Antibody

CCL24/Eotaxin-2/MPIF-2 Monoclonal specifically detects CCL24/Eotaxin-2/MPIF-2 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CCL24/Eotaxin-2/MPIF-2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 61016.11
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6
Gene Symbols CCL24
Host Species Mouse
Immunogen Purified recombinant human eotaxin 2, expressed in E. coli as immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 6369
Test Specificity When used as a capture in human eotaxin 2 sandwich ELISAs, this exhibits less than 0.03 % cross-reactivity with recombinant human eotaxin, MIP1 alpha, and MCP3.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.