Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL24/Eotaxin-2/MPIF-2 Antibody (MM0149-8C72), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB361331 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361331 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361331 Supplier Novus Biologicals Supplier No. NBP212108MFV450

Mouse Monoclonal Antibody

CCL24/Eotaxin-2/MPIF-2 Monoclonal antibody specifically detects CCL24/Eotaxin-2/MPIF-2 in Human samples. It is validated for Western Blot, Neutralization

Specifications

Antigen CCL24/Eotaxin-2/MPIF-2
Applications Western Blot, Neutralization
Classification Monoclonal
Clone MM0149-8C72
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6
Host Species Mouse
Immunogen Human recombinant CCL24/Eotaxin-2/MPIF-2
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 6369
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.