Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL24/Eotaxin-2/MPIF-2 Antibody (RM0176-8M17) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019052 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019052 0.025 mg
NBP212109 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019052 Supplier Novus Biologicals Supplier No. NBP2121090.025MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

CCL24/Eotaxin-2/MPIF-2 Monoclonal specifically detects CCL24/Eotaxin-2/MPIF-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.

Specifications

Antigen CCL24/Eotaxin-2/MPIF-2
Applications Western Blot, Immunohistochemistry, Blocking Assay, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0176-8M17
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Blocking/Neutralizing 50-200 molar excess
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q9JKC0
Gene Alias chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6
Gene Symbols CCL24
Host Species Rat
Immunogen Mouse Eotaxin 2
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 6369
Test Specificity Mouse. Human reactivity reported in scientific literature (PMID: 24080254).
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.