Learn More
Description
Specifications
Specifications
| Antigen | CCL25/TECK |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Functional Assay |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1-0.2 ug/ml, ELISA 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 0.5-1.0 ug/ml, Functional 10.0-15 ug/ml |
| Formulation | Phosphate buffered saline with No Preservative |
| Gene Accession No. | O15444 |
| Gene Alias | C-C motif chemokine 25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine |
| Gene Symbols | CCL25 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
