Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL27/CTACK Antibody (RM0178-6M35), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP212113A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP212113A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP212113A70 Supplier Novus Biologicals Supplier No. NBP212113AF750
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCL27/CTACK Monoclonal specifically detects CCL27/CTACK in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CCL27/CTACK
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0178-6M35
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ALP, C-C motif chemokine 27, chemokine (C-C motif) ligand 27, CTACKSmall-inducible cytokine A27, CTAK, cutaneous T-cell attracting chemokine, Cutaneous T-cell-attracting chemokine, ESKINE, IL-11 Ralpha-locus chemokine, ILCCC chemokine ILC, PESKY, SCYA27IL-11 R-alpha-locus chemokine, skinkine, small inducible cytokine subfamily A (Cys-Cys), member 27
Gene Symbols CCL27
Host Species Rat
Immunogen Mouse recombinant CCL27/CTACK
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10850
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.