Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL3/CCL4 Antibody (93342), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB348810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348810 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348810 Supplier Novus Biologicals Supplier No. FAB2701MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCL3/CCL4 Monoclonal specifically detects CCL3/CCL4 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen CCL3/CCL4
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 93342
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C-C motif chemokine;C-C motif chemokine 3;G0S19-1;LD78ALPHA;MIP1A;MIP-1-alpha;SCYA3
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL3/MIP-1 alpha , Ala27-Ala92, Accession # P10147
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6348
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.