Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL3/MIP-1 alpha Antibody (39624), PerCP, Novus Biologicals™
SDP

Catalog No. NB296256 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296256 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296256 Supplier Novus Biologicals Supplier No. FAB450C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCL3/MIP-1 alpha Monoclonal antibody specifically detects CCL3/MIP-1 alpha in Mouse samples. It is validated for Western Blot

Specifications

Antigen CCL3/MIP-1 alpha
Applications Western Blot
Classification Monoclonal
Clone 39624
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78alpha, Mip1a, MIP-1alpha, MIP1-alpha, Scya3
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL3/MIP-1 alpha , Ala24-Ala92, Accession # P10855
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6348
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.