Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL3L1/LD78 beta Antibody, Novus Biologicals™
SDP

Catalog No. NBP128465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128465 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128465 Supplier Novus Biologicals Supplier No. NBP128465
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CCL3L1/LD78 beta Polyclonal specifically detects CCL3L1/LD78 beta in Human samples. It is validated for Peptide ELISA.

Specifications

Antigen CCL3L1/LD78 beta
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_066286.1
Gene Alias C-C motif chemokine 3-like 1, CCL3L3, chemokine (C-C motif) ligand 3-like 1, D17S1718, G0/G1 switch regulatory protein 19-2, G0S19-2LD78, LD78BETA, LD78-beta(1-70), MGC104178, MGC12815, MGC182017, MIP1AP, PAT 464.2, SCYA3L, SCYA3L1464.2, small inducible cytokine A3-like 1, Small-inducible cytokine A3-like 1, Tonsillar lymphocyte LD78 beta protein
Gene Symbols CCL3L1
Host Species Goat
Immunogen Peptide with sequence CNQVLSAPLAADT corresponding to internal region according to NP_066286.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 6349
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.