Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL4/MIP-1 beta Antibody (24006), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB364430 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364430 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364430 Supplier Novus Biologicals Supplier No. FAB271E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCL4/MIP-1 beta Monoclonal specifically detects CCL4/MIP-1 beta in Human samples. It is validated for CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry (Intracellular Staining), Neutralization, Western Blot.

Specifications

Antigen CCL4/MIP-1 beta
Applications CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry (Intracellular Staining), Neutralization, Western Blot
Classification Monoclonal
Clone 24006
Conjugate DyLight 405
Dilution Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CCL4/MIP-1 beta , Ala24-Asn92, Accession # P13236
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6351
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.