Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL4L1/LAG-1 Antibody (RM0181-5G13), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB361308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361308 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361308 Supplier Novus Biologicals Supplier No. NBP212119MFV450

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

CCL4L1/LAG-1 Monoclonal antibody specifically detects CCL4L1/LAG-1 in Mouse samples. It is validated for ELISA, Neutralization

Specifications

Antigen CCL4L1/LAG-1
Applications ELISA, Neutralization
Classification Monoclonal
Clone RM0181-5G13
Conjugate mFluor Violet 450 SE
Dilution ELISA, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias AT744.2, C-C motif chemokine 4-like, CCL4Lsmall inducible cytokine A4-like, chemokine (C-C motif) ligand 4-like 1, LAG1CCL4L2, LAG-1chemokine (C-C motif) ligand 4-like, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, macrophage inflammatory protein-1b2, MIP-1-beta, Monocyte adherence-induced protein 5-alpha, SCYA4L, SCYA4L1, SCYA4L2, Small-inducible cytokine A4-like
Host Species Rat
Immunogen Mouse recombinant CCL4L1/LAG-1
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 9560
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.