Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL5/RANTES Antibody (21445), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB304280 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304280 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304280 Supplier Novus Biologicals Supplier No. FAB278A

Mouse Monoclonal Antibody

CCL5/RANTES Monoclonal antibody specifically detects CCL5/RANTES in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, Flow Cytometry, CyTOF

Specifications

Antigen CCL5/RANTES
Applications Western Blot, Immunohistochemistry, Neutralization, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 21445
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL5/RANTES
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 6352
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.