Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL5/RANTES Rat, DyLight 755, Clone: 53405.111, Novus Biologicals™
SDP

Catalog No. NB2010394IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB2010394IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB2010394IR Supplier Novus Biologicals Supplier No. NB12010394IR

Rat Monoclonal Antibody

CCL5/RANTES Monoclonal specifically detects CCL5/RANTES in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CCL5/RANTES
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 53405.111
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES
Gene Symbols CCL5
Host Species Rat
Immunogen Recombinant full length protein (Mouse) expressed in E.coli.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 6352
Test Specificity RANTES (53405.111)
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.