Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL7/MCP-3/MARC Antibody (063), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB068966 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB068966 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB068966 Supplier Novus Biologicals Supplier No. NBP290018C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CCL7/MCP-3/MARC Monoclonal specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for ELISA.

Specifications

Antigen CCL7/MCP-3/MARC
Applications ELISA
Classification Monoclonal
Clone 63
Conjugate DyLight 650
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL7/MCP-3/MARC (NP_006264.2; Gln24-Leu99).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6354
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.