Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL8/MCP-2 Antibody (35509.11), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB2010391AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB2010391AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB2010391AI Supplier Novus Biologicals Supplier No. NB12010391AF700

Mouse Monoclonal Antibody

CCL8/MCP-2 Monoclonal specifically detects CCL8/MCP-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CCL8/MCP-2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 35509.11
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8
Gene Symbols CCL8
Host Species Mouse
Immunogen Recombinant human monocyte chemotactic protein 2 (MCP2), expressed in E. coli.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6355
Test Specificity By ELISA, this antibody shows cross-reactivity with recombinant human MCP1 and recombinat mouse MIP1 alpha. In addition, it does not cross-react with other chemokines tested: rmC10, rhENA78, rhGRO alpha, rhGRO beta, rhGRO gamma, rmJE, rhI309, and rhIL8. This antibody neutralizes the biological activity of recombinant human MCP-2. Shows <20% cross-reactivity with recombinant human MCP-1 and recombinant mouse MIP-1a.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.