Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL8/MCP-2 Antibody (35509.11), Novus Biologicals™
SDP

Catalog No. NB12010391 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12010391 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12010391 Supplier Novus Biologicals Supplier No. NB12010391

Mouse Monoclonal Antibody

CCL8/MCP-2 Monoclonal specifically detects CCL8/MCP-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CCL8/MCP-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Neutralization
Classification Monoclonal
Clone 35509.11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, ELISA 1 - 2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen, Neutralization 1 - 4 ug/ml
Gene Alias C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8
Gene Symbols CCL8
Host Species Mouse
Immunogen Recombinant human monocyte chemotactic protein 2 (MCP2), expressed in E. coli.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6355
Test Specificity By ELISA, this antibody shows cross-reactivity with recombinant human MCP1 and recombinat mouse MIP1 alpha. In addition, it does not cross-react with other chemokines tested: rmC10, rhENA78, rhGRO alpha, rhGRO beta, rhGRO gamma, rmJE, rhI309, and rhIL8. This antibody neutralizes the biological activity of recombinant human MCP-2. Shows <20% cross-reactivity with recombinant human MCP-1 and recombinant mouse MIP-1a.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.