Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL9 Antibody (RM0185-28B3) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212408
Description
CCL9 Monoclonal specifically detects CCL9 in Mouse samples. It is validated for Western Blot, Block/Neutralize.Specifications
CCL9 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1:2000, Blocking/Neutralizing 50-200 molar excess | |
P51670 | |
CCL9 | |
Mouse recombinant CCL9 | |
0.1 mg | |
Chemokines and Cytokines | |
20308 | |
Mouse | |
Purified |
Western Blot, Blocking Assay | |
RM0185-28B3 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
CCF18, Ccl9 chemokine (C-C motif) ligand 9, MRP-2, Scya10, Scya9 | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction