Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL9 Antibody (RM0185-28B3) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50
Specifications
Antigen | CCL9 |
---|---|
Clone | RM0185-28B3 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:2000, Blocking/Neutralizing 50-200 molar excess |
Applications | Blocking Assay, Western Blot |
Description
CCL9 Monoclonal specifically detects CCL9 in Mouse samples. It is validated for Western Blot, Block/Neutralize.Specifications
CCL9 | |
LYOPH | |
Blocking Assay, Western Blot | |
Unconjugated | |
Rat | |
Chemokines and Cytokines | |
P51670 | |
20308 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RM0185-28B3 | |
Western Blot 1:500-1:2000, Blocking/Neutralizing 50-200 molar excess | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CCF18, Ccl9 chemokine (C-C motif) ligand 9, MRP-2, Scya10, Scya9 | |
CCL9 | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title