Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL9 Antibody (RM0185-28B3), FITC, Novus Biologicals™
SDP

Catalog No. NB360982 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360982 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360982 Supplier Novus Biologicals Supplier No. NBP212408F

Rat Monoclonal Antibody

CCL9 Monoclonal antibody specifically detects CCL9 in Mouse samples. It is validated for Western Blot, Neutralization

Specifications

Antigen CCL9
Applications Western Blot, Neutralization
Classification Monoclonal
Clone RM0185-28B3
Conjugate FITC
Dilution Western Blot, Block/Neutralize
Formulation PBS
Gene Alias CCF18, Ccl9 chemokine (C-C motif) ligand 9, MRP-2, Scya10, Scya9
Host Species Rat
Immunogen Mouse recombinant CCL9
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 20308
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.