Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCNB1IP1 Antibody (OTI4H3), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N272106J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272106J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272106J646 Supplier Novus Biologicals Supplier No. NBP272106JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCNB1IP1 Monoclonal specifically detects CCNB1IP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen CCNB1IP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI4H3
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias chromosome 14 open reading frame 18, cyclin B1 interacting protein 1, cyclin B1 interacting protein 1, E3 ubiquitin protein ligase, E3 ubiquitin-protein ligase CCNB1IP1, EC 6.3.2.-, enhancer of invasion 10, HEI10C14orf18Cyclin-B1-interacting protein 1, Human enhancer of invasion 10
Gene Symbols CCNB1IP1
Host Species Mouse
Immunogen Full length recombinant protein of human CCNB1IP1 (NP_878269) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 57820.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.