Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCNK Antibody, Novus Biologicals™
SDP

Catalog No. NBP106519 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106519 Supplier Novus Biologicals Supplier No. NBP106519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCNK Polyclonal specifically detects CCNK in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen CCNK
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:1000 - 1:5000, Immunoprecipitation 2 -10 μg/mg lysate, Immunohistochemistry-Paraffin 1:1000 - 1:5000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. O75909
Gene Alias CPR4MGC9113, cyclin K, cyclin-K
Gene Symbols CCNK
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 530 and 580 of human cyclin K using the numbering given in entry NP_001092872.1 (GeneID 8812).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8812
Test Specificity Human. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine, Dog, Guinea pig_10141, Pig, Rhesus Monkey, Chimpanzee, Tasmanian devil, Crab-eating macaque, African elephant, Naked mole rat, Gray short-tailed opossum, Small-eared galago and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.