Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCNY Antibody (OTI12F10), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP272462V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272462V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272462V Supplier Novus Biologicals Supplier No. NBP272462V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCNY Monoclonal specifically detects CCNY in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen CCNY
Applications Western Blot
Classification Monoclonal
Clone OTI12F10
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CBCP1C10orf9cyc-Y, CCNX, CFP 1, CFP1FLJ95513, chromosome 10 open reading frame 9, Cyclin box protein 1, Cyclin fold protein 1, cyclin Y, cyclin-box carrying protein 1, cyclin-X, cyclin-Y, Cyc-Y
Gene Symbols CCNY
Host Species Mouse
Immunogen Full length recombinant protein of human CCNY (NP_859049) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 219771.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.