Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR1 Antibody (003), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB088097 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB088097 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB088097 Supplier Novus Biologicals Supplier No. NBP289235AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CCR1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human CCR1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1230
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.