Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR1 Antibody (643854R), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB311845 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311845 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311845 Supplier Novus Biologicals Supplier No. FAB5986RPECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCR1 Monoclonal specifically detects CCR1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR1
Applications Flow Cytometry
Classification Monoclonal
Clone 643854R
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCR1, Extracellular regions (aa 1-34, aa 92-107, aa 172-192, aa 265-281), Accession # P51675
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1230
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.