Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR10/GPR2 Antibody (314305R) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB372276 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372276 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372276 Supplier Novus Biologicals Supplier No. FAB3478RJ
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCR10/GPR2 Monoclonal antibody specifically detects CCR10/GPR2 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen CCR10/GPR2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 314305R
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2G-protein coupled receptor 2
Host Species Rat
Immunogen Y3 rat myeloid cell line transfected with human CCR10, Gln8-Asn362, Accession # P46092
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2826
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.