Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR10/GPR2 Antibody, Novus Biologicals™
SDP

Catalog No. NB394428 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB394428 25 μL
NBP256544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB394428 Supplier Novus Biologicals Supplier No. NBP25654425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCR10/GPR2 Polyclonal specifically detects CCR10/GPR2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CCR10/GPR2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2G-protein coupled receptor 2
Gene Symbols CCR10
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LRFRQDLRRLLRGGSCPSGPQPRRGCPRRPRLSSCSAPTETHSLSWDN
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2826
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.