Learn More
Description
Specifications
Specifications
| Antigen | CCR10/GPR2 |
| Applications | Immunocytochemistry, Immunofluorescence, Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunohistochemistry, Immunohistochemistry (Frozen), Simple Western, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, Simple Western 1:50, Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunohistochemistry-Frozen 1:10-1:2000. Use reported in scientific literature (Vestergaard et al (2003)), Flow (Cell Surface), Flow (Intracellular) reported in scientific literature (PMID 15663561) |
| Formulation | PBS with 0.2% BSA with 0.05% Sodium Azide |
| Gene Accession No. | P46092 |
| Gene Alias | CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2G-protein coupled receptor 2 |
| Gene Symbols | CCR10 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
