Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR2 Antibody (890231R) [Allophycocyanin], Novus Biologicals™
SDP

Catalog No. NB373029 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373029 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373029 Supplier Novus Biologicals Supplier No. FAB8368RA
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCR2 Monoclonal specifically detects CCR2 in Rat samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CCR2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 890231R
Conjugate APC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with rat CCR2, Accession # O55193
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 729230
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.