Learn More
Description
Specifications
Specifications
| Antigen | CCR2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Accession No. | P41597 |
| Gene Alias | C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor |
| Gene Symbols | CCR2 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
