Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR3 Antibody (5E8-G9-B4), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB014785 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014785 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014785 Supplier Novus Biologicals Supplier No. NBP281046FR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CCR3 Monoclonal specifically detects CCR3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR3
Applications Flow Cytometry
Classification Monoclonal
Clone 5E8-G9-B4
Conjugate DyLight 680
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841
Gene Symbols CCR3
Host Species Rabbit
Immunogen Recombinant monoclonal antibody to CCR3 (CD193).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1232
Test Specificity Recognises human CCR3, a chemokine receptor highly expressed of eosinophils, as well as Th2-Tcells, basophils, platelets, mast-cells, mononuclear phagocytes, airway epithelial cells, and hematopoietic progenitor cells (CD34+).
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.