Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR3 Antibody (61828) [DyLight 550], Novus Biologicals™
SDP

Catalog No. NB399547 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399547 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399547 Supplier Novus Biologicals Supplier No. FAB155L
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCR3 Monoclonal antibody specifically detects CCR3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF

Specifications

Antigen CCR3
Applications Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF
Classification Monoclonal
Clone 61828
Conjugate DyLight 550
Dilution Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841
Host Species Rat
Immunogen Y3 rat myeloid cell line transfected with human CCR3, Accession # P51677
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1232
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.