Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056335 0.1 mg
NB10056335T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056335 Supplier Novus Biologicals Supplier No. NB10056335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

CCR3 Polyclonal specifically detects CCR3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen CCR3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. NP_847899
Gene Alias b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841
Gene Symbols CCR3
Host Species Rabbit
Immunogen This antibody was raised against a synthetic peptide of human CCR3 protein.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1232
Test Specificity In Daudi, a 45 kDa band should be observed.
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.