Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR4 Antibody (MM0064-9G12), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP212129UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212129UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212129UV Supplier Novus Biologicals Supplier No. NBP212129UV

Mouse Monoclonal Antibody

CCR4 Monoclonal specifically detects CCR4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR4
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MM0064-9G12
Conjugate DyLight 350
Dilution Flow Cytometry, Immunohistochemistry-Frozen
Gene Accession No. P51679
Gene Alias C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5
Gene Symbols CCR4
Host Species Mouse
Immunogen Human CCR4 transfected 293 cells
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1233
Test Specificity Detects human CCR4, not other CCRs
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.