Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR4, Rabbit anti-Human, Clone: KW-0761 (Mogamulizumab), Novus Biologicals™
SDP

Catalog No. NB016830 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016830 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016830 Supplier Novus Biologicals Supplier No. NBP2812700.2MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CCR4 Monoclonal specifically detects CCR4 in Human samples. It is validated for Western Blot, Functional.

Specifications

Antigen CCR4
Applications Functional Assay, Depletion Assay
Classification Monoclonal
Clone KW-0761 (Mogamulizumab)
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Functional, Cell depletion
Formulation PBS
Gene Alias C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5
Gene Symbols CCR4
Host Species Rabbit
Immunogen This antibody was prepared by humanization of a mouse anti-CCR4 mAb.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1233
Test Specificity This antibody is specific for human CCR4, a chemokine receptor that is preferentially expressed by Th2 and regulatory T (Treg) cells. CCR4 is also detected on basophils, platelets, monocytes and brain microvascular and coronary artery endothelial cells.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.