Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR5 Antibody (225307), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB367476 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367476 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367476 Supplier Novus Biologicals Supplier No. FAB18041Z
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCR5 Monoclonal specifically detects CCR5 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CCR5
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 225307
Conjugate DyLight 755
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
Host Species Rat
Immunogen C6 rat glioma cell line transfected with mouse CCR5, Accession # P51682
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1234
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.