Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR5 Mouse anti-Human, Clone: E11/19, Novus Biologicals™
SDP

Catalog No. NB152585 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB152585 0.025 mg
NB152586 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB152585 Supplier Novus Biologicals Supplier No. NBP3119940.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunoprecipitation

Specifications

Antigen CCR5
Applications Western Blot, ELISA, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone E11/19
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
Host Species Mouse
Immunogen This antibody was raised by immunizing mice with a synthetic phosphopeptide (CEAPERA(pS)(pS)VYTR(pS)TGEQI(pS)VGL) corresponding to the C-terminal region of human CCR5.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1234
Target Species Human
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.