Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR6 Antibody (140706), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB311876 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311876 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311876 Supplier Novus Biologicals Supplier No. FAB590APCCY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCR6 Monoclonal specifically detects CCR6 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR6
Applications Flow Cytometry
Classification Monoclonal
Clone 140706
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22
Host Species Rat
Immunogen RBL-1 rat basophilic leukemia cell line transfected with mouse CCR6, Met1-Met367, Accession # O54689
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1235
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.