Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR6 Antibody (53103), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB366933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366933 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366933 Supplier Novus Biologicals Supplier No. FAB195M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCR6 Monoclonal specifically detects CCR6 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen CCR6
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 53103
Conjugate DyLight 594
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CCR6, Met1-Met374, Accession # P51684
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1235
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.