Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR6 Antibody (876515), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB346197 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346197 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346197 Supplier Novus Biologicals Supplier No. FAB8320MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCR6 Monoclonal specifically detects CCR6 in Rat samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CCR6
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 876515
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22
Host Species Mouse
Immunogen NSO mouse myeloma cell line transfected with rat CCR6, Met1-Met366, Accession # Q5BK58
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1235
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.